(S)-(+)-Prolinol 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: (S)-(+)-2-Hydroxymethylpyrrolidine, (2S)-pyrrolidin-2-ylmethanol, L(+)-Prolinol, S(+)-2-Pyrrolidinemethanol, L-Prolinol

OR5872-1G OR5872-500G OR5872-100G
77423-256EA 21.26 USD
77423-256 77423-258 77423-254
(S)-(+)-Prolinol 98%
(S)-(+)-Prolinol
Formula: C₅H₁₁NO
MW: 101.15 g/mol
Boiling Pt: 74…76 °C/2 mmHg
Density: 1.025 g/cm³ (25 °C)
Flash Pt: 86 °C
Storage Temperature: Ambient
MDL Number: MFCD00005255
CAS Number: 23356-96-9
EINECS: 245-605-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR