2'-Aminoacetophenone 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR55399-100G OR55399-1KG OR55399-500G
77423-048EA 37.32 USD
77423-048 77423-050 77423-052
2'-Aminoacetophenone 95%
2'-Aminoacetophenone
Formula: C₈H₉NO
Storage Temperature: Refrigerator
MDL Number: MFCD00007717
CAS Number: 551-93-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR