8-Chlorotetrazolo[1,5-a]pyridine 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR6102-100MG OR6102-250MG
77423-454EA 41.78 USD
77423-454 77423-456
8-Chlorotetrazolo[1,5-a]pyridine 98%
8-Chlorotetrazolo[1,5-a]pyridine
Formula: C₅H₃ClN₄
Melting Pt: 194…200 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00490111
CAS Number: 40971-88-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR