3-Benzyl-5-(2-hydroxyethyl)-4-methylthiazolium chloride

Supplier: APOLLO SCIENTIFIC
Warning

OR61012-10G OR61012-5G
77423-444EA 28.52 USD
77423-444 77423-446
3-Benzyl-5-(2-hydroxyethyl)-4-methylthiazolium chloride
3-Benzyl-5-(2-hydroxyethyl)-4-methylthiazolium chloride
Formula: C₁₃H₁₆ClNOS
Melting Pt: 142…144 °C
Storage Temperature: Ambient
MDL Number: MFCD00011959
CAS Number: 4568-71-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR