4-Chloropicolinamide 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Chloropyridine-2-carboxamide

OR5724-1G OR5724-25G OR5724-5G
77423-186EA 21.26 USD
77423-186 77423-188 77423-190
4-Chloropicolinamide 98%
4-Chloropicolinamide
Formula: C₆H₅ClN₂O
MW: 156.57 g/mol
Melting Pt: 160…163 °C
Storage Temperature: Ambient
MDL Number: MFCD01085339
CAS Number: 99586-65-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR