Fmoc-Pro-OH 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR54567-100G OR54567-1KG OR54567-500G
77422-900EA 89.66 USD
77422-900 77422-902 77422-904
Fmoc-Pro-OH 98%
Fmoc-Pro-OH
Formula: C₂₀H₁₉NO₄
Melting Pt: 117…118 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00037122
CAS Number: 71989-31-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR