3,4,5-Trimethoxyaniline 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR7231-100G OR7231-500G OR7231-25G
77424-158EA 20.6 USD
77424-158 77424-156 77424-160
3,4,5-Trimethoxyaniline 98%
3,4,5-Trimethoxyaniline
Formula: C₉H₁₃NO₃
MW: 183.21 g/mol
Melting Pt: 110…114 °C
Storage Temperature: Ambient
MDL Number: MFCD00008393
CAS Number: 24313-88-0
EINECS: 246-154-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR