Triphenylmethylamine 98%

Supplier: APOLLO SCIENTIFIC

OR50478-100G OR50478-1KG OR50478-500G
77422-348EA 96.78 USD
77422-348 77422-350 77422-352
Triphenylmethylamine 98%
Triphenylmethylamine
Formula: C₁₉H₁₇N
Storage Temperature: Ambient
MDL Number: MFCD00008047
CAS Number: 5824-40-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR