2-(1H-Imidazol-5-yl)acetic acid hydrochloride 95%

Supplier: APOLLO SCIENTIFIC

Synonyms: Imidazole-4-acetic acid monohydrochloride4-Imidazoleacetic acid hydrochloride

OR51912-1G
77422-544EA 21.63 USD
77422-544
2-(1H-Imidazol-5-yl)acetic acid hydrochloride 95%
2-(1H-Imidazol-5-yl)acetic acid hydrochloride
Formula: C₅H₇ClN₂O₂
MW: 162.58 g/mol
Melting Pt: 218…222 °C
Storage Temperature: Ambient
MDL Number: MFCD00012698
CAS Number: 3251-69-2
EINECS: 221-840-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR