Dichloro(1,5-cyclooctadiene)palladium(II) 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1,5-Cyclooctadienepalladium(II) Dichloride

OR46568-1G OR46568-5G
77421-866EA 101.88 USD
77421-866 77421-868
Dichloro(1,5-cyclooctadiene)palladium(II) 98%
Dichloro(1,5-cyclooctadiene)palladium(II)
Formula: C₈H₁₂Cl₂Pd
MW: 285.50 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012412
CAS Number: 12107-56-1
EINECS: 235-161-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR