1,3-Diphenylacetone

Supplier: APOLLO SCIENTIFIC

OR52322-100G OR52322-250G OR52322-25G
77422-666EA 63.16 USD
77422-666 77422-668 77422-670
1,3-Diphenylacetone
1,3-Diphenylacetone
Formula: (C₆H₅)CH₂C(O)CH₂(C₆H₅)
Boiling Pt: 330 °C
Melting Pt: 32…34 °C
Density: 1.04 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00004795
CAS Number: 102-04-5
EINECS: 203-000-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR