2-Methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3-thiazole 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Methoxy-1,3-thiazole-5-boronic acid, pinacol ester, 2-Methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiazole

OR46024-1G OR46024-250MG
77421-752EA 508 USD
77421-752 77421-754
2-Methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3-thiazole 95%
2-Methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3-thiazole
Formula: C₁₀H₁₆BNO₃S
Storage Temperature: Freezer
MDL Number: MFCD10688611
CAS Number: 1312765-17-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR