7-Bromoindan-1-one

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 7-Bromo-1-indanone, 7-Bromo-2,3-dihydro-1H-inden-1-one, 7-Bromo-2,3-dihydro-1-oxo-1H-indene

OR40392-1G OR40392-5G
77421-120EA 86.2 USD
77421-120 77421-122
7-Bromoindan-1-one
7-Bromoindan-1-one
Formula: C₉H₇BrO
MW: 211.055 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06796574
CAS Number: 125114-77-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR