6-Bromo-3-chloroisoquinoline 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR40220-1G OR40220-5G
77421-056EA 104.94 USD
77421-056 77421-058
6-Bromo-3-chloroisoquinoline 97%
6-Bromo-3-chloroisoquinoline
Formula: C₉H₅BrClN
Storage Temperature: Refrigerator
MDL Number: MFCD08703290
CAS Number: 552331-06-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR