Ethanesulfonamide 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR51744-10G OR51744-1G OR51744-5G
77422-484EA 28.32 USD
77422-484 77422-486 77422-488
Ethanesulfonamide 98%
Ethanesulfonamide
Formula: C₂H₇NO₂S
MW: 109.1475 g/mol
Melting Pt: 57…60 °C
Storage Temperature: Ambient
MDL Number: MFCD03265410
CAS Number: 1520-70-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR