4-Bromo-3-chlorostyrene 95%

Supplier: APOLLO SCIENTIFIC

OR400918-1G OR400918-250MG
77420-958EA 578.68 USD
77420-958 77420-960
4-Bromo-3-chlorostyrene 95%
4-Bromo-3-chlorostyrene
Formula: C₈H₆BrCl
Storage Temperature: Refrigerator
MDL Number: MFCD30237713
CAS Number: 1692077-74-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR