4-Bromo-2,6-dichloropyridine 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR43622-10G OR43622-1G OR43622-5G
77421-470EA 167.08 USD
77421-470 77421-472 77421-474
4-Bromo-2,6-dichloropyridine 97%
4-Bromo-2,6-dichloropyridine
Formula: C₅H₂BrCl₂N
Melting Pt: 95…102 °C
Storage Temperature: Ambient
MDL Number: MFCD11848646
CAS Number: 98027-80-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR