Iodomethylboronic acid, pinacol ester 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR361281-10G OR361281-1G OR361281-5G
77420-640EA 55.02 USD
77420-640 77420-642 77420-644
Iodomethylboronic acid, pinacol ester 95%
Iodomethylboronic acid, pinacol ester
Formula: C₇H₁₄BIO₂
Storage Temperature: Freezer
MDL Number: MFCD12405515
CAS Number: 70557-99-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR