1,1-Dimethylbiguanide hydrochloride

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: AMPK Signaling Activator VII, Metformin, Metformin hydrochloride, 1,1-Dimethylbiguanide Hydrochloride

OR322044-100G OR322044-10G OR322044-25G
77420-050EA 59.1 USD
77420-050 77420-052 77420-054
1,1-Dimethylbiguanide hydrochloride
1,1-Dimethylbiguanide hydrochloride
Formula: C₄H₁₂ClN₅
Melting Pt: 221 °C
Density: 1.36 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012582
CAS Number: 1115-70-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR