3-Amino-4-isobutoxybenzoic acid

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3-Amino-4-(2-methylpropoxy)benzoic acid, 5-Carboxy-2-isobutoxyaniline

OR40321-500MG OR40321-250MG
77421-076EA 221.08 USD
77421-076 77421-074
3-Amino-4-isobutoxybenzoic acid
3-Amino-4-isobutoxybenzoic acid
Formula: C₁₁H₁₅NO₃
MW: 209.242 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12168706
CAS Number: 1096806-43-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR