(4-Dimethylaminophenyl)di-tert-butylphosphine 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-(Di-tert-butylphosphino)-N,N-dimethylaniline

OR315484-100G OR315484-25G OR315484-5G
77419-604EA 461.52 USD
77419-604 77419-606 77419-608
(4-Dimethylaminophenyl)di-tert-butylphosphine 97%
(4-Dimethylaminophenyl)di-tert-butylphosphine
Formula: C₁₆H₂₈NP
Melting Pt: 57…61 °C
Storage Temperature: Ambient
MDL Number: MFCD09265102
CAS Number: 932710-63-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR