3-Amino-2-pyridone 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3-Amino-2-hydroxypyridine, 3-Amino-2-pyridinol, 3-Amino-2-pyridinone

OR315027-1G OR315027-25G OR315027-5G
77419-528EA 48.9 USD
77419-528 77419-530 77419-532
3-Amino-2-pyridone 98%
3-Amino-2-pyridone
Formula: C₅H₆N₂O
Storage Temperature: Ambient
MDL Number: MFCD03411556
CAS Number: 33630-99-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR