2,2,6,6-Tetramethylpiperidin-4-ol 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2,2,6,6-Tetramethyl-4-piperidinol, 4-Hydroxy-2,2,6,6-tetramethylpiperidine

OR6938-100G OR6938-1KG OR6938-50G
77410-948EA 23.44 USD
77410-948 77410-950 77410-952
2,2,6,6-Tetramethylpiperidin-4-ol 98%
2,2,6,6-Tetramethylpiperidin-4-ol
Formula: C₉H₁₉NO
MW: 157.26 g/mol
Boiling Pt: 212…215 °C
Melting Pt: 129…133 °C
Density: 1.085 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005983
CAS Number: 2403-88-5
EINECS: 219-291-2
UN: 3259
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR