Methyl-4-aminopicolinate 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Aminopyridine-2-carboxylic acid methyll ester, Methyl 4-Aminopyridine-2-carboxylate, 4-Aminopicolinic acid methyll ester, Methyl 4-Aminopicolinate

OR303823-5G OR303823-1G OR303823-25G
77411-316EA 117.16 USD
77411-316 77411-312 77411-314
Methyl-4-aminopicolinate 97%
Methyl 4-aminopicolinate
Formula: C₇H₈N₂O₂
Melting Pt: 129…130 °C
Storage Temperature: Ambient
MDL Number: MFCD00956000
CAS Number: 71469-93-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR