4-Bromo-1-butanol, Technical Grade

Supplier: APOLLO SCIENTIFIC

Synonyms: 1-Bromo-4-hydroxybutane

OR4233-25G OR4233-5G
77409-660EA 166.06 USD
77409-660 77409-662
4-Bromo-1-butanol, Technical Grade
4-Bromo-1-butanol
Formula: BrH₂C(CH₂)₂CH₂OH
MW: 153.02
Boiling Pt: 63…65 °C (5 mmHg)
Density: 1.68 g/cm³
Flash Pt: 7 °C
Storage Temperature: Freezer
MDL Number: MFCD00015387
CAS Number: 33036-62-3
EINECS: 251-355-5
UN: 1987
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR