1-(2-Aminoethyl)pyrrolidine 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: N-(2-Aminoethyl)pyrrolidine, Pyrrolidinoethylamine

OR19557-100G OR19557-25G OR19557-5G
77406-496EA 195.62 USD
77406-496 77406-498 77406-500
1-(2-Aminoethyl)pyrrolidine 98%
1-(2-Aminoethyl)pyrrolidine
Formula: C₆H₁₄N₂
MW: 114.19 g/mol
Boiling Pt: 161 °C
Density: 0.901 g/cm³ (25 °C)
Flash Pt: 47 °C
Storage Temperature: Ambient
MDL Number: MFCD00003182
CAS Number: 7154-73-6
EINECS: 230-509-5
UN: 2734
ADR: 8+3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR