1,10-Phenanthroline 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: o-Phenanthroline

OR2343-100G OR2343-25G OR2343-5G
77407-326EA 66.22 USD
77407-326 77407-328 77407-330
1,10-Phenanthroline 98%
1,10-Phenanthroline
Formula: C₁₂H₈N₂
MW: 180.21 g/mol
Boiling Pt: 360
Melting Pt: 114…120 °C
Storage Temperature: Ambient
MDL Number: MFCD00011678
CAS Number: 66-71-7
EINECS: 200-629-2
UN: 2811
ADR: 6.1 III
Merck Index: 12,07356

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR