1,1,1-Trifluoro-2-propanol 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC7386-100G PC7386-25G
77417-850EA 544.04 USD
77417-850 77417-852
1,1,1-Trifluoro-2-propanol 97%
1,1,1-Trifluoro-2-propanol
Formula: C₃H₅F₃O
Boiling Pt: 75…76 °C
Density: 1.259 g/cm³ (25 °C)
Flash Pt: 18 °C
Storage Temperature: Ambient
MDL Number: MFCD00014403
CAS Number: 374-01-6
EINECS: 206-773-2
UN: 1987
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR