Trifluoromethanesulfonic anhydride 99%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
Synonyms:
Triflic anhydride
PC7420-1KG
PC7420-250G
PC7420-500G
77417-866EA
368.82
USD
77417-866
77417-868
77417-870
Trifluoromethanesulfonic anhydride 99%
Trifluoromethanesulphonic anhydride
Formula:
C₂F₆O₅S₂ Boiling Pt: 81…83 °C Melting Pt: –80 °C Density: 1.677 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000408 CAS Number: 358-23-6 EINECS: 206-616-8 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...