Potassium hexafluorophosphate 99%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
PC6290-100G
PC6290-1KG
PC6290-250G
77417-356EA
63.16
USD
77417-356
77417-358
77417-360
Potassium hexafluorophosphate 99%
Potassium hexafluorophosphate
Soluble neutral salt that can be used for synthesis of other hexafluorophosphate salts as a substitutefor hexafluorophosphoric acid.
Formula:
F₆KP Melting Pt: 575 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011412 CAS Number: 17084-13-8 EINECS: 241-143-0 UN: 3260 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...