5-Amino-2,2-difluoro-1,3-benzodioxole 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2,2-Difluoro-1,3-benzodioxol-5-amine, 3,4-(Difluoromethylenedioxy)aniline

PC3252-5G PC3252-10G PC3252-1G
77414-614EA 23.28 USD
77414-614 77414-610 77414-612
5-Amino-2,2-difluoro-1,3-benzodioxole 98%
5-Amino-2,2-difluoro-1,3-benzodioxole
Formula: C₇H₅F₂NO₂
MW: 173.12 g/mol
Boiling Pt: 119
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00190144
CAS Number: 1544-85-0
UN: 2810
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR