4-Fluoro-3-methylbenzylamine 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC0357-100G PC0357-25G PC0357-5G
77412-508EA 163.52 USD
77412-508 77412-510 77412-512
4-Fluoro-3-methylbenzylamine 97%
4-Fluoro-3-methylbenzylamine
Formula: C₈H₁₀FN
Boiling Pt: 145 °C
Density: 1.059 g/cm³ (25 °C)
Flash Pt: 68 °C
Storage Temperature: Ambient
MDL Number: MFCD01631490
CAS Number: 261951-68-2
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR