α-Bromo-3-chloro-5-fluorotoluene 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 1-(Bromomethyl)-3-chloro-5-fluorobenzene, 3-Chloro-5-fluorobenzyl bromide

PC0198-1G PC0198-25G PC0198-5G
77412-456EA 21.26 USD
77412-456 77412-458 77412-460
α-Bromo-3-chloro-5-fluorotoluene 98%
α-Bromo-3-chloro-5-fluorotoluene
Formula: C₇H₅BrClF
Storage Temperature: Ambient
MDL Number: MFCD03788472
CAS Number: 493024-39-8
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR