4-Amino-1-boc-piperidine 97%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
Synonyms:
tert-Butyl 4-Amino-1-piperidinecarboxylate, 4-Amino-1-tert-butoxycarbonylpiperidine, tert-Butyl 4-aminopiperidine-1-carboxylate, N-Boc-4-piperidineamine, tert-Butyl 4-amino-1-piperidinecarboxylate, 4-Amino-1-Boc-piperidine, 4-Amino-1-piperidinecarboxylic acid tert-Butyl Ester
OR9845-100G
OR9845-25G
OR9845-5G
77412-298EA
69.28
USD
77412-298
77412-300
77412-302
4-Amino-1-boc-piperidine 97%
4-Amino-1-Boc-piperidine
Formula:
C₁₀H₂₀N₂O₂ MW: 200.28 g/mol Boiling Pt: 80 °C/0.037 mmHg Melting Pt: 44…48 °C Storage Temperature: Ambient |
MDL Number:
MFCD01076201 CAS Number: 87120-72-7 UN: 3259 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...