Benzoyl chloride 99%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Benzoic acid chloride

OR5454-250G OR5454-1KG
77410-394EA 83.54 USD
77410-394 77410-392
Benzoyl chloride 99%
Benzoyl chloride
Formula: C₇H₅ClO
MW: 140.57 g/mol
Boiling Pt: 197.2
Melting Pt: -0.6
Density: 1.211 g/cm³ (25 °C)
Flash Pt: 72 °C
Storage Temperature: Ambient
MDL Number: MFCD00000653
CAS Number: 98-88-4
EINECS: 202-710-8
UN: 1736
ADR: 8 II
Merck Index: 13,01113

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR