2,6-Lutidine 98+%

Supplier: APOLLO SCIENTIFIC
Warning

OR5334-1KG OR5334-250G
77410-294EA 140.6 USD
77410-294 77410-296
2,6-Lutidine 98+%
2,6-Lutidine
Formula: C₇H₉N
Boiling Pt: 143…145 °C
Melting Pt: –6 °C
Density: 0.921 g/cm³ (25 °C)
Flash Pt: 33 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006345
CAS Number: 108-48-5
EINECS: 203-587-3
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR