3,4-Diethoxy-3-cyclobutene-1,2-dione 98%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
OR26825-5G
OR26825-25G
77407-854EA
21.34
USD
77407-854
77407-852
3,4-Diethoxy-3-cyclobutene-1,2-dione 98%
3,4-Diethoxy-3-cyclobutene-1,2-dione
Formula:
C₈H₁₀O₄ Boiling Pt: 94…95 °C/0.1 mmHg Density: 1.15 g/cm³ (25 °C) Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001333 CAS Number: 5231-87-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...