2-Methylimidazole 98%

Supplier: APOLLO SCIENTIFIC
Danger

OR2900-100G OR2900-25G OR2900-500G
77408-312EA 36.68 USD
77408-312 77408-314 77408-316
2-Methylimidazole 98%
2-Methylimidazole
Formula: C₄H₆N₂
Boiling Pt: 267…268 °C
Melting Pt: 142…144 °C
Flash Pt: 155 °C
Storage Temperature: Ambient
MDL Number: MFCD00005190
CAS Number: 693-98-1
EINECS: 211-765-7
UN: 1759
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR