2-Amino-5-methylpyridine 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 6-Amino-3-picoline, 5-Methyl-2-pyridylamine, 5-Methylpyridin-2-amine, 2-Amino-5-picoline

OR28779-100G OR28779-500G OR28779-50G
77408-228EA 28.52 USD
77408-228 77408-230 77408-232
2-Amino-5-methylpyridine 98%
2-Amino-5-methylpyridine
Formula: C₆H₈N₂
Boiling Pt: 226…228 °C
Melting Pt: 74…78 °C
Flash Pt: 118 °C
Storage Temperature: Ambient
MDL Number: MFCD00006328
CAS Number: 1603-41-4
EINECS: 216-503-5
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR