Bromotrimethylsilane
Supplier: APOLLO SCIENTIFIC
|
Danger
|
OR2120-100G
OR2120-25G
OR2120-500G
77406-634EA
36.68
USD
77406-634
77406-636
77406-638
Bromotrimethylsilane
Bromotrimethylsilane
Formula:
C₃H₉BrSi Boiling Pt: 79 °C Melting Pt: –43 °C Density: 1.17 g/cm³ (25 °C) Flash Pt: –8 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00000048 CAS Number: 2857-97-8 EINECS: 220-672-0 UN: 2924 ADR: 3+8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...