Tributylallyltin

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Allyltributylstannane, Allyltributyltin, Tributylallylstannane, Tributyl-2-propenylstannane, Allyltri-n-butyltin, Tri-n-butyl-2-propenylstannane, Allyltri-n-butylstannane

OR16616-100G OR16616-25G
77405-768EA 338.24 USD
77405-768 77405-770
Tributylallyltin
Tributylallyltin
Formula: C₁₅H₃₂Sn
MW: 331.13 g/mol
Boiling Pt: 88
Density: 1.068 g/cm³ (25 °C)
Flash Pt: 103 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00010346
CAS Number: 24850-33-7
EINECS: 246-494-3
UN: 2788
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR