4-tert-Butylbenzenesulfonyl chloride

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 4-tert-Butylbenzene-1-sulfonyl chloride, 4-tert-Butylbenzene-1-sulphonyl chloride

OR1355-25G OR1355-5G
77404-666EA 48.64 USD
77404-666 77404-668
4-tert-Butylbenzenesulfonyl chloride
4-tert-Butylbenzenesulphonyl chloride
Formula: C₁₀H₁₃ClO₂S
Melting Pt: 78…81 °C
Storage Temperature: Ambient
MDL Number: MFCD00007449
CAS Number: 15084-51-2
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR