1,4-Dihydro-2,3-quinoxalinedione-2-oxime 95%

Supplier: APOLLO SCIENTIFIC

OR12080-10G OR12080-1G OR12080-5G
77404-152EA 564.42 USD
77404-152 77404-154 77404-156
1,4-Dihydro-2,3-quinoxalinedione-2-oxime 95%
1,4-Dihydro-2,3-quinoxalinedione-2-oxime
Formula: C₈H₇N₃O₂
Melting Pt: 262…264 °C
Storage Temperature: Ambient
MDL Number: MFCD08689712
CAS Number: 177944-77-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR