18-Crown-6 (1,4,7,10,13,16-hexaoxacyclooctadecane) 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR10919-100G OR10919-25G OR10919-500G
77402-974EA 23.42 USD
77402-974 77402-976 77402-978
18-Crown-6 (1,4,7,10,13,16-hexaoxacyclooctadecane) 98%
18-Crown-6 (1,4,7,10,13,16-Hexaoxacyclooctadecane)
Formula: C₁₂H₂₄O₆
Boiling Pt: 116 °C/0.2 mmHg
Melting Pt: 37…45 °C
Density: 1.175 g/cm³ (25 °C)
Flash Pt: 109 °C
Storage Temperature: Ambient
MDL Number: MFCD00005113
CAS Number: 17455-13-9
EINECS: 241-473-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR