L(+)-Cysteine
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR0325-100G
OR0325-250G
77402-020EA
29
USD
77402-020
77402-022
77402-024
L(+)-Cysteine
L-Cysteine
Formula:
HSCH₂CH(NH₂)CO₂H Melting Pt: 240 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00064306 CAS Number: 52-90-4 EINECS: 200-158-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...