3-Aminobenzenesulfonamide

Supplier: APOLLO SCIENTIFIC
Warning

OR017076-100G OR017076-25G
77401-808EA 132.44 USD
77401-808 77401-810
3-Aminobenzenesulfonamide
3-Aminobenzenesulphonamide
Formula: C₆H₈N₂O₂S
MW: 172.21 g/mol
Melting Pt: 139…142 °C
Storage Temperature: Ambient
MDL Number: MFCD00035781
CAS Number: 98-18-0
EINECS: 202-646-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR