Cerium trichloride hydrate 99,9%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Cerium(III) chloride hydrateCerous chloride

IN1414-250G
77401-390EA 216 USD
77401-390
Cerium trichloride hydrate 99,9%
Cerium trichloride hydrate
Formula: CeCl₃·xH₂O
MW: 246.48 g/mol (anhy)
Melting Pt: 848 °C
Storage Temperature: Ambient
MDL Number: MFCD00149633
CAS Number: 19423-76-8
EINECS: 232-227-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR