Boron phosphate 99,9%

Supplier: APOLLO SCIENTIFIC
Warning

IN1296-100G
77401-380EA 288.4 USD
77401-380
Boron phosphate 99,9%
Boron phosphate
Formula: BO₄P
MW: 105.78 g/mol
Melting Pt: >1400 °C
Density: 2.81 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011318
CAS Number: 13308-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR