2-Ethynylaniline 95%

Supplier: APOLLO SCIENTIFIC

Synonyms: 2-Aminophenylacetylene

OR015038-1G OR015038-10G OR015038-5G
77401-740EA 77.25 USD
77401-740 77401-742 77401-738
2-Ethynylaniline 95%
2-Ethynylaniline
Formula: C₈H₇N
MW: 117.15 g/mol
Boiling Pt: 229…230 °C
Density: 1.03 g/cm³ (25 °C)
Flash Pt: 98.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00168862
CAS Number: 52670-38-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR