Magnesium chloride, anhydrous 99%

Supplier: APOLLO SCIENTIFIC
Warning

IN1017-100G IN1017-1KG IN1017-500G
77401-348EA 35.68 USD
77401-348 77401-350 77401-352
Magnesium chloride, anhydrous 99%
Magnesium chloride
Formula: MgCl₂
MW: 95.21 g/mol
Boiling Pt: 1412
Melting Pt: 715
Density: 2.32 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011106
CAS Number: 7786-30-3
EINECS: 232-094-6
Merck Index: 13,05684

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR